Cholesterol cell-based detection assay kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10009779-192
CAYM10009779-192EA 374 EUR InStock
CAYM10009779-192
Cholesterol cell-based detection assay kit
Tests Zelltests
Cholesterol Cell-Based Detection Assay Kit is a 192-well assay for measurement of cholesterol.
  • Detect cholesterol uptake and localisation within cultured cells
  • Employs filipin III, a probe that fluoresces in the presence of cholesterol
  • An inhibitor of cholesterol trafficking is included as a positive control
The mechanism for the movement of cholesterol from intracellular sites to their ultimate cellular destination is an unresolved question of fundamental importance to cell biology and medicine. Thus, defining mechanisms of intracellular cholesterol transport and identifying the cellular factors involved are therefore of great interest. The Cholesterol Cell-Based Detection Assay Kit provides a simple fluorometric method to study mechanisms and biological factors that regulate cholesterol metabolism or movement within cells.
Lieferumfang: The kit includes filipin III, fixative, and wash buffer in a ready to use format.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..