Triglyceride colorimetric assay kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10010303-96
CAYM10010303-96EA 317 EUR InStock
CAYM10010303-96
Triglyceride colorimetric assay kit
Tests Zelltests
Triglyceride Colorimetric Assay Kit is a simple, reproducible and sensitive tool for assaying triglycerides in plasma, serum, cell lysates and tissue homogenate samples.
  • Measure TG levels in plasma, serum, cell lysates, and tissue homogenates
  • Assay 26 samples in triplicate and 40 samples in duplicate
  • Assay range: 3,125 - 200 mg/dl
  • Incubation: 15 minutes
  • Read: Colorimetric at 530 - 550 nm
The measurement of triglyceride levels, in conjunction with other lipid assays, are useful in the diagnosis of primary and secondary hyperlipoproteinemia, dyslipidemia, and triglyceridemia. This kit provides a simple, reproducible, and sensitive tool for assaying triglycerides in plasma, serum, cell lysates, and tissue homogenate samples. The assay is initiated with the enzymatic hydrolysis of the triglycerides by lipase to produce glycerol and free fatty acids. The glycerol released is subsequently measured by a coupled enzymatic reaction system with a colorimetric readout at 540 nm.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge