Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Lutein ≥98%, kristalliner Feststoff

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

10010811-1 10010811-5 10010811-10
CAYM10010811-1EA 50 EUR InStock
CAYM10010811-1 CAYM10010811-5 CAYM10010811-10
Lutein ≥98%, kristalliner Feststoff
Lutein
Lutein (Xanthophyll) is a natural yellow carotenoid, which can be found in plants, egg yolks, and in the human retina. It is a lipid-soluble antioxidant that can circulate in the plasma and confer cardioprotective, anti-inflammatory, and anti-angiogenic effects. Dietary lutein, in combination with zeaxanthin, vitamins and ω-3 fatty acids, may be beneficial against age-related macular degeneration and cataract development.


Formel: C₄₀H₅₆O₂
Storage Temperature: Ultra-Tiefkühlschrank (–80°C)
MDL Number: MFCD08435941
CAS-Nummer: 127-40-2
EINECS: 204-840-0
Order Now
Spezifikation Testergebnisse
Purity ≥98%
Formulation A crystalline solid
λmax 268, 445, 474 nm
Storage – 80 °C
Stability ≥2 years

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge