Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

2,3',4,5'-Tetramethoxystilbene ≥98%

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

10038-1 10038-10 10038-5 10038-50
CAYM10038-1EA 22 EUR InStock
CAYM10038-1 CAYM10038-10 CAYM10038-5 CAYM10038-50
2,3',4,5'-Tetramethoxystilbene ≥98%
2,3',4,5'-Tetramethoxystilbene
TMS is a potent and selective inhibitor of CYP1B1, with an IC50 of 6 nM.

CYP1B1 is mainly an extrahepatic enzyme which oxidatively metabolises both endogenous (steroids; eicosanoids) and exogenous xenobiotics such aspolyaromatic hydrocarbons.

It is 50-fold selective for the inhibition of CYP1B1 versus CYP1A1, making it a useful tool to differentiate between various CYP450 families. In cultured human colon cancer cells, TMS induces apoptosis and inhibits cell growth with an IC50 of 0.8 µg/ml.


Formel: C₁₈H₂₀O₄
Melting Pt: -99 °C
Storage Temperature: Gefrierschrank
MDL Number: MFCD03428621
CAS-Nummer: 24144-92-1
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge