Prostaglandin Screening Library II
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10502-50
CAYM10502-50EA 1552 EUR InStock
CAYM10502-50
Prostaglandin Screening Library II
Tests Zelltests
For D and E series prostaglandin analog screening.
  • Contains >60 compounds
  • Includes the ''D- and E-series'' prostaglandins
The Prostaglandin Screening Library II contains more than 60 ''D and E-series'' of prostaglandins in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge