Fatty Acid Screening Library
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10504-50
CAYM10504-50EA 1958 EUR InStock
CAYM10504-50
Fatty Acid Screening Library
Tests Zelltests
For screening of variety of fatty acids.
  • Contains >75 compounds
  • Includes arachidonic, linoleic, eicosapentaenoic, stearic, α- and γ-linolenic, oleic, and palmitic acids
The Fatty Acid Screening Library contains more than 75 compounds of fatty acids with diverse biological activities in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The library may also include some of the most commonly-ordered fatty acids, including arachidonic, linoleic, eicosapentaenoic, stearic, α- and γ-linolenic, oleic, and palmitic acids. As well as some unusual fatty acids, including 9-thiastearic acid, 2-hydroxymyristic acid, AUDA, and CUDA may be included. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge