Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine ≥98%

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


10621-25 10621-5 10621-100 10621-10
CAYM10621-25EA 190 EUR InStock
CAYM10621-25 CAYM10621-5 CAYM10621-100 CAYM10621-10
1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine ≥98%
1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine
Olaparib is a potent inhibitor of PARP1 and PARP2 (IC50 = 5 and 1 nM, respectively) but is less effective against the PARP tankyrase-1 (IC50 = 1,5 µM). It can be used in cells and in animals, alone or in combination therapy with alkylating agents, to block BER and increase cancer cell death.

These products generated by alkylating agents on DNA can be efficiently repaired by normal base excision repair (BER). Some poly(ADP-ribose) polymerases (PARPs) assist in the repair of single-strand DNA nicks, an important step in BER.


Formel: C₂₄H₂₃FN₄O₃
MDL Number: MFCD13185161
CAS-Nummer: 763113-22-0
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge