Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

1,1-Diphenyl-2-(2,4,6-trinitrophenyl)hydrazin ≥95%, kristalliner Feststoff

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

14805-50 14805-100 14805-500
CAYM14805-50EA 53 EUR InStock
CAYM14805-50 CAYM14805-100 CAYM14805-500
1,1-Diphenyl-2-(2,4,6-trinitrophenyl)hydrazin ≥95%, kristalliner Feststoff
2,2-Diphenyl-1-picrylhydrazyl (free radical)
DPPH, known formally as 2,2-diphenyl-1-picrylhydrazyl, is a cell-permeable, stable free radical that is commonly used to evaluate the ability of compounds to act as free radical scavengers or hydrogen donors and to measure the antioxidant activity of tissue extracts.

  • Synonyms: 2,2-Diphenyl-1-Picrylhydrazyl; DPPH radical

The reaction of DPPH with an antioxidant or reducing compound produces the corresponding hydrazine DPPH2, which can be followed by color change from purple (absorbance at 515 to 528 nm) to yellow.

DPPH is sparingly soluble in aqueous solutions. To enhance aqueous solubility, dilute the organic solvent solution into aqueous buffers or isotonic saline. If performing biological experiments, ensure the residual amount of organic solvent is insignificant, since organic solvents may have physiological effects at low concentrations. Not recommend storing the aqueous solution for more than one day.


Formel: C₁₈H₁₂N₅O₆
Storage Temperature: Kühlschrank
MDL Number: MFCD00007231
CAS-Nummer: 1898-66-4
EINECS: 217-591-8
Order Now
Spezifikation Testergebnisse
Purity ≥95%
Stability ≥2 years at −20 °C
Form Crystalline Solid
UV/Vis λmax: 327, 515 nm

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge