Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Busulfan ≥98%, kristalliner Feststoff

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

14843-25 14843-50 14843-100 14843-250
CAYM14843-25EA 65 EUR InStock
CAYM14843-25 CAYM14843-50 CAYM14843-100 CAYM14843-250
Busulfan ≥98%, kristalliner Feststoff
Busulphan
1,4-Butanediol Dimethanesulfonate (Busulfan) is an alkyl sulfonate that acts as an alkylating antineoplastic agent. It forms both intra- and interstrand crosslinks on DNA. In mammals, busulfan causes profound and prolonged reduction in the generation of hematopoietic progenitors without significantly affecting lymphocyte levels and humoral antibody responses.

  • Soluble in DMSO and dimethylformamide
  • Stable for atleast two years

Synonyms: Busulphan, Mielosan, Milecitan, Myeloleukon, Mylecytan, Myleran, NCI C01592, NSC 750


Formel: C₆H₁₄O₆S₂
MW: 246.29 g/mol
Melting Pt: 114…118°C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00007562
CAS-Nummer: 55-98-1
UN: 2811
ADR: 6.1 I
Order Now
Spezifikation Testergebnisse
Purity ≥98%
Stability ≥2 years at −20 °C
Form Crystalline solid
Solubility Sparingly soluble in aqueous buffers
Storage temperature −20 °C

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge