Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Calcein fluoreszierender Indikator

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

16221-1 16221-25 16221-5 16221-10
CAYM16221-1EA 43 EUR InStock
CAYM16221-1 CAYM16221-25 CAYM16221-5 CAYM16221-10
Calcein fluoreszierender Indikator
Calcein
Calcein is a xanthene that is commonly used for the fluorometric determination of calcium in solution. As this form of calcein is not membrane permeable, it can be used in assays that evaluate membrane integrity. Calcein can also be used as a fluorescent indicator for fluoride, iron, and mercury. Excitation and emission maxima for calcein are 494 and 517 nm, respectively.

  • Emission 517 nm
  • Excitation 494 nm
  • Formulation A crystalline solid
  • λmax 226, 278 nm

This form of calcein is not membrane permeable and can be used in assays that evaluate membrane integrity.


Formel: C₃₀H₂₆N₂O₁₃
Storage Temperature: Gefrierschrank
MDL Number: MFCD00005049
CAS-Nummer: 1461-15-0
EINECS: 215-957-1
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge