Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Luminol (3-Aminophthalhydrazid) ≥98%

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

16803-10 16803-25 16803-5 16803-50
CAYM16803-10EA 104 EUR InStock
CAYM16803-10 CAYM16803-25 CAYM16803-5 CAYM16803-50
Luminol (3-Aminophthalhydrazid) ≥98%
Luminol (3-Aminophthalhydrazid)
Luminol is a cyclic diacyl hydrazide that exhibits chemiluminescence upon oxidation.

  • Emission 411 nm
  • Excitation 355 nm
  • Formulation A crystalline solid
  • λmax 220, 295, 355 nm

HRP combined with hydrogen peroxide as an activator causes luminescent peroxidation of luminol. This reaction can be enhanced by certain phenol derivatives, such as p-substituted phenols. Luminol can also be oxidised and chemiluminesce by compounds containing iron, copper, gold and cyanide.


Formel: C₈H₇N₃O₂
MW: 177.16 g/mol
Storage Temperature: Gefrierschrank
MDL Number: MFCD00006890
CAS-Nummer: 521-31-3
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge