Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Tenofovirdisoproxilfumarat ≥98%, kristalliner Feststoff

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

16922-10 16922-25 16922-5 16922-50
CAYM16922-10EA 145 EUR InStock
CAYM16922-10 CAYM16922-25 CAYM16922-5 CAYM16922-50
Tenofovirdisoproxilfumarat ≥98%, kristalliner Feststoff
Tenofovir disoproxil fumarate
Tenofovir disoproxil is a prodrug of the acyclic nucleoside phosphonate, tenofovir. Tenofovir is converted by cellular enzymes to tenofovir diphosphate, an obligate chain terminator that inhibits the activity of HIV reverse transcriptase and hepatitis B virus polymerase. Due to its rapid intracellular uptake, the anti-HIV activity of tenofovir disoproxil is reportedly >100-fold greater than that of the negatively charged tenofovir in a T cell line and primary blood lymphocytes.

  • Crystalline solid
  • Soluble in organic solvents such as ethanol, DMSO and dimethyl formamide (DMF)
  • Sparingly soluble in aqueous buffers


Formel: C₁₉H₃₀N₅O₁₀P·C₄H₄O₄
Storage Temperature: Gefrierschrank
MDL Number: MFCD08141829
CAS-Nummer: 202138-50-9
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge