Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Biotin hydrazide ≥95%

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


18418-1 18418-100 18418-250 18418-500
CAYM18418-1EA 438 EUR InStock
CAYM18418-1 CAYM18418-100 CAYM18418-250 CAYM18418-500
Biotin hydrazide ≥95%
Biotin hydrazide
Biotin hydrazide is a biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups.

Used as a probe for determining protein carbonylation, an irreversible posttranslational modification resulting from the actions of reactive oxygen species or lipid oxidation products. The reaction of this probe is direct and does not require catalysts or reducing agents.


Formel: C₁₀H₁₈N₄O₂S
MDL Number: MFCD00078532
CAS-Nummer: 66640-86-6
Order Now
Spezifikation Testergebnisse
Purity ≥95%
Formulation A Crystalline Solid

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge