Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sinensetin ≥98%

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

19751-10 19751-25 19751-5
CAYM19751-10EA 223 EUR InStock
CAYM19751-10 CAYM19751-25 CAYM19751-5
Sinensetin ≥98%
Sinensetin
Sinensetin is a polymethoxylated flavone that is present in the Southeast Asian medical plant O. stamineus, as well as in orange oil. It has been shown to enhance adipogenesis and lipolysis by increasing cAMP levels in 3T3-L1 cells. Sinensetin is also reported to inhibit α-glucosidase and α-amylase activity in vitro, indicating its potential usefulness in the control of glucose absorption. Also it is used to suppress the expression of genes associated with inflammation by regulating IκBα protein levels in LPS-activated macrophages.


Formel: C₂₀H₂₀O₇
MW: 372.4 g/mol
Storage Temperature: Gefrierschrank
MDL Number: MFCD00017421
CAS-Nummer: 2306-27-6
UN: 2811
ADR: 6.1 III
Order Now
Spezifikation Testergebnisse
Purity ≥98%
Formulation A Crystalline Solid
λmax 215, 241, 327 nm

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge