Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sisomicin ≥98%

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

21046-1 21046-100 21046-50 21046-500
CAYM21046-1EA 405 EUR InStock
CAYM21046-1 CAYM21046-100 CAYM21046-50 CAYM21046-500
Sisomicin ≥98%
Sisomicin
Sisomicin is a broad-spectrum aminoglycoside antibiotic originally isolated from M. inyoensis.

It is active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa among others (MICs = 0.01-0.75 µg/ml) but is less active against P. multocida and V. coli (MICs = 7.5 and 2.4 µg/ml, respectively). Sisomicin is protective against Gram-positive and Gram-negative bacterial infections in mice with 50% protective dose (PD50s) values ranging from 0.5 to 6 mg/kg.


Formel: C₁₉H₃₇N₅O₇
MW: 447.50 g/mol
Storage Temperature: Gefrierschrank
MDL Number: MFCD00865123
CAS-Nummer: 32385-11-8
EINECS: 251-018-2
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge