Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cysteaminiumchlorid ≥95%, kristalliner Feststoff

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

22193-10 22193-100 22193-25 22193-50
CAYM22193-10EA 43 EUR InStock
CAYM22193-10 CAYM22193-100 CAYM22193-25 CAYM22193-50
Cysteaminiumchlorid ≥95%, kristalliner Feststoff
Cysteaminiumchlorid
Cysteamine is a stable aminothiol with radioprotective activities.

It reduces ionizing radiation-induced death and chromosomal damage in mice in a dose-dependent manner. Cysteamine binds rapidly and temporarily to plasma proteins upon administration and this activity is directly correlated to its radioprotective effects. In vitro, 0.1 mM cysteamine depletes 90% of free cystine from cystinotic fibroblasts. Formulations containing cysteamine have been used to treat nephropathic cystinosis and reduce glomerular deterioration in humans.

Achtung! Bitte beachten: This product is not for human or veterinary use.


Formel: C₂H₇NS·HCl
Storage Temperature: Gefrierschrank
MDL Number: MFCD00012904
CAS-Nummer: 156-57-0
UN: 3335
ADR: 9,III
Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge