DHA Oxylipin LC-MS Mixture, Analytical Standard
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
22280-1
CAYM22280-1EA 483 EUR InStock
CAYM22280-1
DHA Oxylipin LC-MS Mixture, Analytical Standard
Standards Organische Standards
The DHA oxylipin LC-MS mixture contains oxylipin metabolites derived from docosahexaenoic acid.
  • Each vial contains a mixture of standards packaged in amber ampules purged with argon
  • >1 ml provided at the concentration specified in the insert
  • Designed for direct snap and inject use in mass spectrometry applications
  • Ideally suited for method development and as a system suitability standard
The mixture is supplied in an amber ampule in which the headspace has been purged with argon to prevent lipid oxidation. This product has been designed for direct use in LC-MS applications. The solution may be used as a system suitability standard or tuning standard. After opening, we recommend that the mixture be transferred immediately to a 1 ml glass screw cap vial, to prevent solvent evaporation, and stored at -20°C. The mixture should be discarded after multiple freeze/thaw cycles. This mixture contains (±)4-HDHA (Item No. 33200), (±)7-HDHA (Item No. 33300), (±)8-HDHA (Item No. 33350), (±)10-HDHA (Item No. 33400), (±)11-HDHA (Item No. 33450), (±)13-HDHA (Item No. 33500), 14(S)-HDHA (Item No. 15253), (±)16-HDHA (Item No. 33600), 17(S)-HDHA (Item No. 10009799), and (±)20-HDHA (Item No. 33750).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..