LIBX-A401 Formal Name: methyl (E)-3-(4-(2-(N-methylbenzamido)ethoxy)phenyl)acrylate Molecular Formula: C20H21NO4 Purity: 95% Storage: -20C Shipping: Room temperature in continental US may vary elsewhere Stability : 2 years 1 * 5 mg
Lieferant: CAYMAN
LIBX-A401 Formal Name: methyl (E)-3-(4-(2-(N-methylbenzamido)ethoxy)phenyl)acrylate Molecular Formula: C20H21NO4 Purity: 95% Storage: -20C Shipping: Room temperature in continental US may vary elsewhere Stability : 2 years 1 * 5 mg
New Product
PRODUKT
Erweitert / Kompakt
LIBX-A401 Formal Name: methyl (E)-3-(4-(2-(N-methylbenzamido)ethoxy)phenyl)acrylate Molecular Formula: C20H21NO4 Purity: ~95% Storage: -20°C Shipping: Room temperature in continental US may vary elsewhere Stability : ~2 years 1 * 5 mg CAYM41744-5 1 * 5 mg
New Product
Hersteller-Artikelnummer:: 41744-5
Lokale Artikelnummer:: CAYM41744-5
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder