500370-96S
500370-96
500370-480S
500370-480
CAYM500370-96SEA
337
EUR
InStock
CAYM500370-96S
CAYM500370-96
CAYM500370-480S
CAYM500370-480
Cortisol express ELISA kit
Tests
ELISAs
Cortisol Express ELISA kit offers a fast, convenient assay format for measurement of cortisol.
- Measure cortisol in urine, plasma, and other sample matrices
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure cortisol levels down to 110 pg/ml
- Rapid assay; get results in under four hours
Cortisol Express ELISA is a competitive assay that permits the rapid measurement of cortisol from biological samples, requiring only a two-hour incubation and one-hour development times. This ELISA offers the convenience of a fast assay while maintaining sensitivity.
Verpackung: Kit contains monoclonal antibody, AChE tracer, standard, ELISA and wash buffer, polysorbate 20, coated plate, cover sheet, ellman's reagent, tracer and antiserum dye.
Achtung! Bitte beachten: Due to an approximate 15% cross-reactivity with dexamethasone, this kit may not be suitable for use with samples that contain high concentrations of dexamethasone, such as those collected during a dexamethasone suppression test.