Cortisol express ELISA kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
500370-96S 500370-96 500370-480S 500370-480
CAYM500370-96SEA 337 EUR InStock
CAYM500370-96S CAYM500370-96 CAYM500370-480S CAYM500370-480
Cortisol express ELISA kit
Tests ELISAs
Cortisol Express ELISA kit offers a fast, convenient assay format for measurement of cortisol.
  • Measure cortisol in urine, plasma, and other sample matrices
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure cortisol levels down to 110 pg/ml
  • Rapid assay; get results in under four hours
Cortisol Express ELISA is a competitive assay that permits the rapid measurement of cortisol from biological samples, requiring only a two-hour incubation and one-hour development times. This ELISA offers the convenience of a fast assay while maintaining sensitivity.
Verpackung: Kit contains monoclonal antibody, AChE tracer, standard, ELISA and wash buffer, polysorbate 20, coated plate, cover sheet, ellman's reagent, tracer and antiserum dye.
Achtung! Bitte beachten: Due to an approximate 15% cross-reactivity with dexamethasone, this kit may not be suitable for use with samples that contain high concentrations of dexamethasone, such as those collected during a dexamethasone suppression test.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge