ELISA Kit, Human Therapeutic IgG1
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
500910-96
CAYM500910-96EA 886 EUR InStock
CAYM500910-96
ELISA Kit, Human Therapeutic IgG1
Tests ELISAs
A generic assay for the quantitation of therapeutic human IgG1 antibodies in monkey serum or plasma.
  • Specifically detect human IgG1 antibodies in monkey plasma or serum
  • Not cross reactive to rhesus or cynomolgus immunoglobulins
  • Useful for pharmacokinetic analysis of therapeutic human IgG₁ antibodies in monkeys
  • Useful for detecting human IgG1 in mouse or rat plasma or serum
Human therapeutic antibodies have become increasingly common components of early drug discovery and development portfolios in the pharmaceutical and biotech industries. As part of preclinical toxicology assessment of these agents, they are routinely tested in non-human primates, primarily in rhesus or cynomolgus monkeys. In order to assess the pharmacokinetics of the human antibodies in monkey serum, it is necessary to have an assay that is capable of distinguishing the experimentally-introduced human IgG from the endogenous monkey IgG. Cayman’s Human Therapeutic IgG1 ELISA kit is an immunometric assay which can be used to measure human IgG₁ in monkey or rodent plasma and serum without prior sample purification. The standard curve spans the range of 3,3 to 200 ng/ml, with an LLOQ of 3,13 ng/ml. The assay has been validated in serum from cynomolgus monkeys.
Lieferumfang: Kit contains: standard, HRP-conjugate, buffers, reagents, and precoated 96-well plate.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge