Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cyclic AMP Select ELISA Kit

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


501040-480 501040-96
CAYM501040-480EA 1722 EUR InStock
CAYM501040-480 CAYM501040-96
Cyclic AMP Select ELISA Kit
Tests ELISAs
For direct quantification of cAMP from a variety of biological samples.

  • A sensitive assay for cAMP in multiple sample matricies without the need for acetylation
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure cAMP levels down to 0,6 pmol/ml
  • Incubation: 18 hours | Development: 90 to 120 minutes | Read: Colorimetric at 405 tp 420 nm

Cayman’s cAMP Select assay is a competitive ELISA that permits cAMP measurements within the standard curve range of 0,09 to 200 pmol/ml and a sensitivity (80% B/B0) of approximately 0,6 pmol/ml. Acetylation of samples is not required in this highly sensitive assay where culture media, urine, tissue, and plasma samples have been validated. Cyclic AMP is a ubiquitous cellular second messenger that is a critical component of a signal transduction pathway linking membrane receptors and their ligands to the activation of internal cellular enzymatic activity and gene expression. cAMP is synthesized from ATP by membrane-bound adenylate cyclase. Binding of certain ligands or hormones to their specific G-protein couple receptors activates GTP binding proteins (Gs or Gi) which either stimulate or inhibit adenylate cyclase. cAMP activates or inhibits various enzymes or cascade of enzymes by promoting their phosphorylation or dephosphorylation. The cAMP signal is neutralized by hydrolysis of cAMP to AMP by phosphodiesterases. Therefore, the concentration of cAMP in a cell is a function of the ratio of the rate of synthesis from ATP by adenylate cyclase and its rate of breakdown to AMP by specific phosphodiesterases.

Lieferumfang: Kit contains: Antiserum, tracer, standard, buffers, reagents, and plate(s).


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge