Vitamin D ELISA kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
501050-480 501050-96 501050-480S 501050-96S
CAYM501050-480EA 2319 EUR InStock
CAYM501050-480 CAYM501050-96 CAYM501050-480S CAYM501050-96S
Vitamin D ELISA kit
Tests ELISAs
Sensitive ELISA for vitamin D quantification in plasma or serum.
  • Measure vitamin D in plasma or serum
  • Detect the metabolically stable forms 25(OH)D₃ and 25(OH)D₂
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure vitamin D levels down to 0,5 ng/ml
  • Read: Colorimetric at 405 to 420 nm
The primary synthesis of functional vitamin D begins in the skin, where a cholesterol by-product acted upon by UV light forms vitamin D₃. Vitamin D₃ is carried through the bloodstream, attached to the vitamin D binding protein, to the liver where it is converted to 25-hydroxy vitamin D₃ (25(OH)D₃). This product is then transported to the kidney where it is further converted to 1,25-dihydroxycholecalciferol (1,25(OH)₂D₃), the biologically active form. Active 1,25(OH)₂D₃ is very short-lived and is rapidly metabolized to its deactivated forms.
Vitamin D Immunoassay, accurate Vitamin D measurement in plasma or serum. Vitamin D ELISA Kit is a competitive assay that can be used for Vitamin D quantification in plasma or serum. Because of the short half-life of the biologically active 1,25(OH)D form, this Vitamin D assay primarily detects the more metabolically stable forms, 25(OH)D₃ and 25(OH)D₂.
Accurate detection of these forms requires that they be displaced from the vitamin D binding protein prior to measurement. A simple, acetone-based purification protocol is included as the initial procedure of the Vitamin D assay kit for this purpose.
Verpackung: Kit contains: Antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, tracer and antiserum dye.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge