Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Leukotriene E4 ELISA Kit

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


501060-480 501060-96
CAYM501060-480EA 1863 EUR InStock
CAYM501060-480 CAYM501060-96
Leukotriene E4 ELISA Kit
Tests ELISAs
For the quantification of LTE4 in urine.

  • Measure LTE4 in urine, with no purification required
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure LTE4 levels down to 25 pg/ml
  • Incubation: 18 hours, Development: 90 to 120 minutes, Read: Colorimetric at 405 to 420 nm

Leukotriene E4 (LTE4) is a product of the 5-lipoxygenase (5-LO) pathway in activated mast cells, eosinophils, and monocytes. LTA4, the primary 5-LO metabolite, is rapidly converted to LTC4 and sequentially to LTD4 and LTE4. While plasma levels of LTC4 are virtually undetectable, exogenously administered LTC4 is recovered in the urine as LTE4 (5 to 13%) and two prominent oxidized metabolites resulting from several cycles of β-oxidation. Cayman’s LTE4 ELISA Kit is a competitive assay developed for the measurement of LTE4 from urine without a purification step. The assay has a range of 7,8 to 1000 pg/ml and a sensitivity (80% B/B0) of approximately 25 pg/ml. This ‘dilute and go’ format has been used successfully to demonstrate significant and reproducible changes in LTE4 levels in the urine of asthmatic patients after antigen challenge. If your samples require additional purification prior to analysis, Cayman’s Cysteinyl Leukotriene Affinity Sorbent can be used for this purpose.

Contents: Antiserum, Tracer, Standard, Buffer, Polysorbate 20, Reagent, Dyes, Plate, and Cover Sheet


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge