15(S)-HETE ELISA kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
534721-480 534721-96 534721-96S 534721-480S
CAYM534721-480EA 1573 EUR InStock
CAYM534721-480 CAYM534721-96 CAYM534721-96S CAYM534721-480S
15(S)-HETE ELISA kit
Tests ELISAs
15(S)-HETE ELISA kit is a competitive assay for the measurement of 15(S)-HETE.
  • Detect 15(S)-HETE, a product of 15-lipoxygenase treatment of arachidonic acid
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 15(S)-HETE levels down to 170 pg/ml
  • Read: Colorimetric at 405 to 420 nm
15(S)-HETE is produced from arachidonic acid by the enzyme 15-LO. In humans it is formed primarily in the respiratory epithelium, leukocytes, and reticulocytes. It may contribute to allergic rhinitis, has anti-inflammatory properties, and acts as a vasoconstrictor.
This assay is very specific for 15(S)-HETE, showing very low crossreactivity to other HETEs.
Lieferumfang: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover sheet, ellman's reagent, tracer and antiserum dye.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge