2-Methoxyestradiol ELISA kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
582261-96S 582261-96 582261-480S 582261-480
CAYM582261-96SEA 615 EUR InStock
CAYM582261-96S CAYM582261-96 CAYM582261-480S CAYM582261-480
2-Methoxyestradiol ELISA kit
Tests ELISAs
A competitive assay that can be used for quantification of 2-methoxyestradiol in plasma, urine, and other sample matrices.
  • Detect 2-ME2
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 2-ME2 levels down to 40 pg/ml
  • Colorimetric read at 405 to 420 nm
2-Methoxyestradiol (2-ME2) is a natural metabolite of estradiol produced by CYP450-dependant hydroxylation followed by catechol-O-methyltransferase-catalyzed O-methylation. Deficiency in catechol-O-methyltransferase is associated with pre-eclampsia. Therefore, 2-ME2 may be utilized as a plasma and urinary diagnostic marker for this disease, as well as a therapeutic supplement for its prevention. Several clinical trials have demonstrated the antitumor effects of 2-ME2 in prostate cancer, breast cancer, renal cell carcinoma, ovarian cancer, glioblastoma, and multiple myeloma. Other studies indicate that 2-ME2 exhibits more potent cardioprotective effects than estradiol. Cayman’s 2-Methoxyestradiol ELISA Kit is a competitive assay that can be used for quantification of 2-ME2 in plasma, urine, and other sample matrices.
Lieferumfang: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover sheet, Ellman's reagent, tracer and antiserum dye.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge