Oct4 transcription factor assay kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
601080-96
CAYM601080-96EA 654 EUR InStock
CAYM601080-96
Oct4 transcription factor assay kit
Tests Zelltests
96-Well assay for measurement of Oct4.
  • A sensitive, non-radioactive method of detecting Oct4 from whole cell lysates
  • 96-well plate format replaces EMSAs
  • Capture the transcription factor using a specific dsDNA sequence bound to the plate
  • Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format
  • Nuclear extraction kit is available to aid in the isolation of nuclear and cytoplasmic fractions from cell lysates or tissue homogenates
Octamer-binding transcription factor 4 (Oct4) is an essential regulator of embryonic stem cell pluripotency and used as a marker for undifferentiated cells. It also plays a role in gene expression, DNA repair and/or replication, signal transduction and tumorigenesis. Cayman’s Oct4 Transcription Factor Assay is a non-radioactive, sensitive method for detecting specific transcription factor-DNA binding activity in nuclear extracts. A 96-well enzyme-linked immunosorbent assay (ELISA) replaces the cumbersome radioactive electrophoretic mobility shift assay (EMSA). A specific double-stranded DNA (dsDNA) sequence containing the Oct4 response element is immobilized onto the wells of a 96-well plate. Oct4, contained in a nuclear extract, binds specifically to the Oct4 response element. Oct4 is detected by the addition of a specific primary antibody directed against Oct4. A secondary antibody conjugated to HRP is added to provide a sensitive colorimetric readout at 450 nm.
Lieferumfang: kit contains assay buffer, reagent A, positive control, binding buffer, primary antibody, wash buffer concentrate, polysorbate 20, competitor dsDNA, HRP conjugate, plate and cover sheet, developing and stop solution.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge