Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

PtdIns-(4,5)-P2 (1,2-dioctanoyl) (sodium salt) ≥97%

Lieferant: CAYMAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


64910-1 64910-5 64910-100 64910-500
CAYM64910-1EA 511 EUR InStock
CAYM64910-1 CAYM64910-5 CAYM64910-100 CAYM64910-500
PtdIns-(4,5)-P2 (1,2-dioctanoyl) (sodium salt) ≥97%
PtdIns-(4,5)-P2 (1,2-dioctanoyl) (sodium salt)
The phosphatidylinositol (PtdIns) represent a small percentage of total membrane phospholipids. However, they play a critical role in the generation and transmission of cellular signals.PtdIns is phosphorylated to mono- (PtdIns-P; PIP), di- (PtdIns-P2; PIP2) and triphosphates (PtdIns-P3; PIP3). Hydrolysis of PIP2 by PI-specific phospholipase C generates IP3 and diacylglycerol (DAG). The DAG and IP3 produced by this reaction are part of a complex biochemical and signal transduction cascade that is at the frontier of scientific research today.

  • Formulation: A lyophilized powder
  • Storage: -20°C
  • Stability: ≥ 1 year


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge