Free Fatty Acid Fluorometric Assay Kit
Lieferant: CAYMAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
700310-96
CAYM700310-96EA 549 EUR InStock
CAYM700310-96
Free Fatty Acid Fluorometric Assay Kit
Tests Enzymtests
A sensitive tool for measuring free fatty acids in plasma, serum, and urine.
  • Measure FFAs in plasma, serum, and urine
  • Assay 20 samples in duplicate
  • Measure free fatty acids from 25 to 250 µM
  • Plate-based fluorometric measurement (ex 530 to 540 nm, em 585 to 595 nm)
The measurement of FFA can be useful in determining metabolic status. Cayman’s Free Fatty Acid (FFA) Fluorometric Assay provides a simple, reproducible, and sensitive tool for measuring free fatty acids in plasma, serum, and urine. The FFA Fluorometric Assay utilizes a coupled enzymatic reaction that results in generation of the highly fluorescent product resorufin.
Lieferumfang: Kit contains: Standard, Fluorometric detector, Assay buffers, Reagents, and 96-Well Plate.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge