Microwave drying oven, SAM-255
Lieferant: CEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
920085
CEMA920085EA 0 EUR InStock
CEMA920085
Microwave drying oven, SAM-255
Öfen Mikrowellen-Trockenschränke
SAM-255 with IntelliTemp™ infrared temperature control provides rapid drying of all types of samples. Workstation can analyse large sized samples for moisture and solids.
  • Intellitemp provides consistent drying every day, every batch
  • Works great with paint bases and colors
  • Ideal for powdered milk
  • Stores up to 20 separate methods and each method can have up to 8 heating, cooling or heat ramping stages
Time and power selections are independently programmed for each stage. The powerful, variable-speed exhaust system moves air through the cavity quickly to aid in drying and can be connected to an existing laboratory exhaust when drying organic compounds.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge