Microwave ovens, Phoenix
Lieferant: CEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S-924560
CEMAS-924560EA 0 EUR InStock
CEMAS-924560
Microwave ovens, Phoenix
Öfen Mikrowellen-Trockenschränke
This rapid microwave ashing system consists of a microwave heating system containing a 5 L chamber which can be heated to 1000 °C or a 1,8 L chamber which can be heated to 1200 °C using microwave energy.
  • Internal temperature calibration software
  • Integral dual thermocouple Temperature Control System for either single set-point temperature or eight segmented ramping temperature control programs
  • Two RS232C 9-pin serial interfaces for option balance and external computer and one 25-pin parallel port for optional printer
The unit maintains 2,8 m³ of air flow per minute through the microwave cavity to rapidly ash materials and remove combustion by-products. Program storing, processing and reporting weight data and % Ash, % LOI, % ROI, % LOD, %Fixed, %Volatile and %Total Residue to the instrument display, printer or external computer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge