Welding face shield
Lieferant: CENTURION SAFETY PRODUCTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S903NCE
CENUS903NCEEA 73.19 EUR InStock
CENUS903NCE
Welding face shield
Gesichtsschutz
This lightweight helmet-mounted welding shield is highly impact resistant.
  • Shield is made of robust Polyamide PA6 (Nylon)
  • Supplied with Polycarbonate chipping lens
  • Compatible with all Centurion helmets except for the Spartan helmet
Window: 110×90 mm
Zertifizierungen: EN 175
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge