Face shields, chin guard system, Connect®
Lieferant: CENTURION SAFETY PRODUCTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S910 S911 S912
CENUS910EA 21.86 EUR InStock
CENUS910 111-1580 CENUS912
Face shields, chin guard system, Connect®
Gesichtsschutz
These clear face shields are available with acetate or PC screens.
  • Chinguard screens have holes at bottom in order to attach chinguard
  • Can be used in combination with a browguard when a safety helmet is not required
Zertifizierungen: EN 166 B
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge