Winkler Electrodes
Lieferant: CHEMPUR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WI1 WI1B
CHMPWI1EA 0 EUR InStock
CHMPWI1 CHMPWI1B
Winkler Electrodes
Elektroden Elektroden, sonstige
Electrode for electrogravimetrical identification of Cu and Zn by Winkler method.
  • Made of platinum/iridium 90/10
This electrode consists of a slotted net cylinder in which a with spiral is concentrically arranged.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge