ST162
YE492
CP572
AQ056
BC732
CQ382
CQ392
EB042
EC168
EC169-25
EE222
EE223-25
EF322
EF323-25
EF342
EL382
LK970
LM852
MC122
MS25P
MS25N
PF652
PS832
RR702
RR703-25
SA162
SA163-25
SQ001
TX182
VB912
VB913-25
CP573-25
SU702
SU703-25
ST162EA
654
EUR
InStock
ST162
YE492
CP572
AQ056
BC732
CQ382
CQ392
EB042
EC168
EC169-25
EE222
EE223-25
EF322
EF323-25
EF342
EL382
LK970
LM852
MC122
MS25P
MS25N
PF652
PS832
RR702
RR703-25
SA162
SA163-25
SQ001
TX182
VB912
VB913-25
CP573-25
CHRASU702
CHRASU703-25
Chromogenic media for colourful microbial detection, CHROMagar™
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
Chromogenic technology applied to culture media allows you to differentiate and easily identify, inside one single plate, the growth of different microorganisms.
- Flexibility of use: Prepare only the quantity you need
- Very easy to prepare
- Long shelf life: 18 months
- Cost saving for overall analysis
- No need for pH adjustment
- Supplements always included in the pack
Dr A. Rambach, founder of CHROMagar™, developed the use of this technology in food microbiology. The main advantage of using chromogenic media is the ability to distinguish with the naked eye different microorganisms growing on the same plate.
All CHROMagar™ products are dehydrated culture media. The unit size of every packs is indicated in volume (litres or millilitres) that shows the volume of culture medium that you can prepare: With one bottle you can prepare 5 or 25 L of culture media.
After the preparation of the medium starting from the powder, the 5 L pack allows you to prepare around 250 plates, while the 25 L allows you to prepare 1250 plates.
Every chromogen exhibits its own distinctive colour; the result is a very specific and distinctive colouring of different microorganisms which become clearly distinguishable to the naked eye under normal lighting conditions.
The 'Rambach Agar', available in this product range, is still one of the most used and famous chromogenic culture media for detection and isolation of Salmonella spp.
Lieferumfang: Supplements always included in the pack.
Verpackung: 5 L pack = dehydrated culture media to prepare 5 L of agar or broth or 25 L pack = dehydrated culture media to prepare 25 L of agar or broth.