,,4 components: Vinylchloride [CAS:75-01-4] 100mg/l Acetaldehyde [CAS:75-07-0] 250mg/l 2-Propanol [CAS:67-63-0] 250mg/l Vinylacetate [CAS:108-05-4] 250mg/l in N,N-Dimethylacetamide ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 6 months.,,,, 1 * 1 SET
Lieferant: CUSTOM MADE CHEMICALS LAB
,,4 components: Vinylchloride [CAS:75-01-4] 100mg/l Acetaldehyde [CAS:75-07-0] 250mg/l 2-Propanol [CAS:67-63-0] 250mg/l Vinylacetate [CAS:108-05-4] 250mg/l in N,N-Dimethylacetamide ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 6 months.,,,, 1 * 1 SET
New Product
PRODUKT
Erweitert / Kompakt
,,4 components: Vinylchloride [CAS:75-01-4] 100mg/l Acetaldehyde [CAS:75-07-0] 250mg/l 2-Propanol [CAS:67-63-0] 250mg/l Vinylacetate [CAS:108-05-4] 250mg/l in N,N-Dimethylacetamide ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 6 months.,,,, 1 * 1 SET CMPLRD1114141.5 1 * 1 SET
New Product
Hersteller-Artikelnummer:: RD1114141.5
Lokale Artikelnummer:: CMPLRD1114141.5
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder