Needle-Free Fluid Transfer System, CODAN Spikes / MicroSpikes
Lieferant: CODAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
165260 16.5292 16.5264 165274 79.3800 79.3801
CODA165260EA 3770 EUR InStock
CODA165260 CODA16.5292 CODA16.5264 CODA165274 CODA79.3800 CODA79.3801
Needle-Free Fluid Transfer System, CODAN Spikes / MicroSpikes
Kanülen
The CODAN Spikes are a compact and safe needle-free system for the manual preparation, withdrawal or transfer of cytostatics and pharmaceuticals as well as their solvent solutions. In order to protect users from aerosols and the drug from micro bacterial contamination, all CODAN Spikes are equipped with a hydrophobic 0,2 µm air filter.
  • Gamma sterilised
  • Cap protects against damage and contamination
  • Integrated, hydrophobic 0,2 µm air filter guarantees sterile aeration and venting and reliably retains aerosols during preparation
  • Minimise the risk of injury
  • Maximise the operating speed by work simplification
The CODAN MicroSpikes were especially designed for use on very small medication vials, e.g. preparation of vaccine.
For the filtration of large particles (e.g. plug punching, glass splinters) CODAN offers Chemoprotect® Spikes with an additional 5 µm fluid filter.
CODAN Spikes / MicroSpikes are also available with SWAN-LOCK® (SWabable Adapter Needle-free - Luer-Lock) access for multi-dose applications.
The handy Chemoprotect® bag with a safe-seal closure is ideal for the transport of medication to the ward and is also excellent for the disposal of substances.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge