Temperature compensator (ATC)
Lieferant: CONSORT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ST10N
CONSST10NEA 135 EUR InStock
CONSST10N
Temperature compensator (ATC)
Temperaturfühler
General purpose temperatur sensor for field use.
  • Stainless steel body, PTFE coated
  • Not suitable for very aggressive fluids
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge