Adaptor, QuickConnect™
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2758
CONT2758EA 79 EUR InStock
CONT2758
Adaptor, QuickConnect™
Mopps
Stainless steel adaptor for use with QuickTask™ mops.
  • Autoclavable at 121 °C for 30 minutes
The QuickConnect™ adaptor allows users to connect any QuickConnect™ handle to the QuickTask™ polypropylene mop frame.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge