Bucket system, 20 L
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2769-KIT
CONT2769-KITEA 1390 EUR InStock
CONT2769-KIT
Bucket system, 20 L
Mopps
Polypropylene bucket system is a stainless steel rolling cart with two polypropylene 20 L buckets, stainless steel drip cap, and angled stainless steel wringer.
  • The red and grey double bucket system allows for differentiation between cleaning and rinsing solutions
  • Compatible with all Contec® flat mops, including our PREMIRA™ advanced environmental hygiene system and Klean Max™
Durable, yet lightweight construction with four standard casters makes it easy to manoeuvre around pharmacy cleanrooms.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge