Cleanroom wipes, Polywipe Light
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7070-6025
CONT7070-6025EA 626 EUR InStock
CONT7070-6025
Cleanroom wipes, Polywipe Light
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
Dry wipes designed for durability and ease of use that are ideal for cage and equipment wipe down or downtime 'super cleaning'.
  • Polyester interlock knit
  • Knife-cut edge
  • Product is double bagged to ensure product integrity
  • Lot numbers provide traceability
  • Economical bulk packaging
The Polywipe Light wipe is an unlaundered standardweight polyester interlock knit wipe with knife-cut edges that is to be used in less than critical applications (in environments of ISO Class 6 and higher). The wipes have the same basic functional characteristics as laundered wipes. Ideal for product or equipment wipe down, downtime clean up and surface preparation, and back-end manufacturing in the electronics industry.
Suitable for use in Class 1000, ISO 6, M4.5 cleanrooms.
Colour: White
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge