Reinraum-Wischtücher, MicroGenesis™
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LWMM0100
CONTLWMM0100EA 995 EUR InStock
CONTLWMM0100
Reinraum-Wischtücher, MicroGenesis™
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
This microfiber wipe is made of 100% polyester microfibre yarn knitted into a thermally bonded PP substrate.
  • Extremely low particle generation
  • Soft texture, performs well wet and dry
  • Silicone-free
  • Safe for use with all disinfectants including bleach and quaternary ammonium
Designed with a micro-terry pile for fast and efficient pick-up and retention of particles, dust, grease, oil, and other contaminants in cleanrooms and other critical applications. The special structure applies microfibre only to the wiping surface where it is needed while sealing the base of each stitch in a thermally bonded base.
Farbe: Weiß.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge