Cleaning swabs, sealed foam, CONSTIX® SF-11
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SF-12
CONTSF-12EA 120 EUR InStock
CONTSF-12
Cleaning swabs, sealed foam, CONSTIX® SF-11
Tupfer Reinigungstupfer
The SF-12 sealed foam swab consists of polyurethane foam bonded to a PP handle without the use of adhesives or binders.
  • Small head size enables good access to small features and difficult to reach surfaces
  • Polyurethane open-cell foam head provides good particle entrapment and removal
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily absorbed for applications involving solvent delivery or removal. The SF-12 swab is excellent for applying resins and adhesives.
Though compatible with many common solvents, the SF-12 is not recommended for use with ketones such as acetone or MEK.
Verpackung: Swabs are packed 500 per case.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge