Cleaning swabs, sealed foam, CONSTIX® SF-13
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SF-13
CONTSF-13EA 175 EUR InStock
CONTSF-13
Cleaning swabs, sealed foam, CONSTIX® SF-13
Tupfer Reinigungstupfer
The SF-13 swab consists of 100 PPI open-cell foam tightly sealed around the tip of a glass-filled PP handle.
  • Polyurethane foam head provides good particle entrapment and removal
  • Foam is secured to the handle without the use of adhesives or binders
  • Very low in particles and fibers
The foam head conforms tightly to the pointed shape of the swab handle to provide a very fine tip that enables access to small areas and crevices. The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal.
Though compatible with many common solvents, the SF-13 is not recommended for use with ketones such as acetone or MEK.
Verpackung: Swabs are packaged 500 per case.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge