Cleaning swabs, sealed foam, CONSTIX® SF-19
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SF-19
CONTSF-19EA 227 EUR InStock
CONTSF-19
Cleaning swabs, sealed foam, CONSTIX® SF-19
Tupfer Reinigungstupfer
The SF-19 sealed foam swab consists of a 100 PPI sealed closed-cell polyurethane foam head formed over the cotton bud of a 6” applicator with wooden handle.
  • Soft polyurethane foam head provides good particle entrapment and removal
  • Wooden handle is solvent resistant and stiff enough to allow vigorous swabbing
  • No adhesives are used
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily absorbed for applications involving solvent delivery and removal. The SF-19 swab is very low in particles and fibres, as well suited to applying solutions and adhesives.
The SF-19 is compatible with most solvents. However, the swab is not recommended for use with ketones such as acetone or MEK.
Verpackung: Swabs are packed 500 per case.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge