Cleaning swabs, sealed foam, CONSTIX® SF-2
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SF-2
CONTSF-2EA 244 EUR InStock
CONTSF-2
Cleaning swabs, sealed foam, CONSTIX® SF-2
Tupfer Reinigungstupfer
The SF-2 swab consists of 100 pore/inch sealed closed-cell foam head with a PP handle.
  • Swab head is thermally sealed and bonded to the handle without adhesives or binders
  • Large rectangular head is ideal for swabbing larger areas and applying coatings
The polyurethane foam head provides good particle entrapment and removal. The large size of the foam head provides better surface coverage for cleaning.
The SF-2 swab is compatible with many solvents, such as isopropanol and ethanol, but is not recommended for use with ketones, such as acetone or MEK.
Verpackung: Swabs are packed 500 per case.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge