Cleaning swabs, sealed, CONSTIX® SP-10
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP-10
CONTSP-10EA 151 EUR InStock
CONTSP-10
Cleaning swabs, sealed, CONSTIX® SP-10
Tupfer Reinigungstupfer
CONSTIX® SP-10 sealed polyester swab consists of 100% knitted polyester fabric that is thermally bonded to a flex-tip PP handle.
  • Double-layer, sealed, knitted polyester swab
  • Polyester head provides enhanced sorbency, particle entrapment and removal
  • Polyester head is compatible with most common solvents, such as alcohols and ketones
The swab head is made of cleanroom laundered; double-layer knitted polyester material. The polyester head is thermally sealed and attached, also constructed without adhesives or binders. The polypropylene handle has a thin, flexible tip that enables access to very small areas and components. The swab is durable, absorbent, and features extremely low particle and residue levels.
Verpackung: Swabs are packed 500 per case.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge