Cleaning swabs, sealed, CONSTIX® SP-6
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP-6
CONTSP-6EA 282 EUR InStock
CONTSP-6
Cleaning swabs, sealed, CONSTIX® SP-6
Tupfer Reinigungstupfer
The SP-6 sealed polyester swab consists of 100% knit polyester fabric that is thermally bonded to a flex-tip PP handle.
  • Flexible tip provides a softer application on surfaces
  • 100% polyester head provides enhanced sorbency, particle entrapment, and removal
  • Head is compatible with most common solvents, such as alcohols and ketones
  • Durable and absorbent
  • Extremely low particles and residue levels
The swab head is made of cleanroom laundered; double-layer knitted polyester. The polyester head is constructed without adhesives or binders.
Verpackung: 500 per case.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge