Polypropylene Wipes, SATWipes®
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SWPP0004
CONTSWPP0004EA 749 EUR InStock
CONTSWPP0004
Polypropylene Wipes, SATWipes®
Wischtücher Reinraum-Wischtücher Getränkte Reinraumwischtücher
Presaturated with IPA/DI water.
  • Rolls of wipes conveniently dispensed through a canister
  • 0.2 micron (µm) filtered provides a safe, consistent, and mild cleaning solution
  • Eliminates the need for solvent bottles to reduce solvent usage and reduces solvent use by up to 35%
SATWIPES® Polypropylene Wipes consist of a roll of (150) 152×230mm saturated wipes dispensed through a center-pull canister with a pop-up dispenser. The wipes are made of soft and clean non-woven polypropylene fabric. The solution is filtered to 0.2µm and provides a safe, consistent, and mild cleaning solution.
Eliminates the need for solvent bottles, reduces solvent handling and reduces solvent use by up to 35%. Refill rolls are available so the dispenser canister can be reused.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge