Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

4N6FLOQ Swabs® Genetics

Lieferant: COPAN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

4520CS01 4508C 4504C 4520CA 40U022D
COPA4520CS01EA 1250 EUR InStock
COPA4520CS01 COPA4508C COPA4504C COPA4520CA COPA40U022D
4N6FLOQ Swabs® Genetics
Tupfer Tupfer für die Probenahme
4N6FLOQ Swabs® Genetics is the line of Copan patented FLOQSwabs® specifically designed and dedicated to DNA collection for human identification.

  • 4N6FLOQ Swabs® optimize the efficiency of the target collection
  • Stability over time
  • Different shapes for different collection
  • Forensic DNA grade
  • FLOQ® technology
  • Breaking point: 20 mm

For more than ten years, the 4N6 FLOQSwabs® Genetics line led the development of a new quality standard for forensics applications. This series delivers a first-rate product line for the whole range of forensic applications, such as databasing, sexual assault, paternity testing, evidence collection, genotyping, and skin microbiome. The active drying system absorbs water molecules, preventing microbial growth and guaranteeing DNA stability for 12+ months at RT and −80 °C.

Shaft/head: Regular or Subungual

Zertifizierungen: ISO 18385:2016. The validated EO treatment assesses the degradation of possible human DNA residuals. Free of amplifiable human DNA, detectable and amplifiable DNase, and RNase. Personnel STR database.

Achtung! Bitte beachten: 4N6FLQ Swabs® Genetics are not intended for diagnostic purposes.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge