Flocked swabs
Lieferant: COPAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
961C 961CS01
COPA961CEA 889 EUR InStock
COPA961C COPA961CS01
Flocked swabs
Tupfer Tupfer für die Probenahme
An extensive range of ready-to-use environmental swabs. The swabls are designed for use in a broad range of environments, including food production, pharmaceutical sites, as well as hospital wards, to determine the microbial bioburden.
  • Sterile options available
  • Varied packaging options
The nylon-flocked swab is designed to improve sample absorption by strong capillary action and to release more than 80% of the collected sample. This increases the efficiency in detecting microorganisms. Traditionally wrapped fiber swabs (rayon or polyester) trap in the fiber matrix a large percentage of the sample, releasing only 20% of it.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge